Learn More
Thermo Scientific™ Oxoid™ Fosfomycin Antimicrobial Susceptibility Disks, 200 μg
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing disks.
Brand: Thermo Scientific™ CT0758B
22.87 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 50
Additional Details : Weight : 1.00000kg
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Fosfomycin Antimicrobial Susceptibility Disks.
The disc contains 200μg of the active ingredient, fosfomycin, with the addition of 50μg of glucose-6-phospate, as required by the EUCAST methodology. Discs must be used for E.coli only and for other Enterobacterales and Staphylococcus spp., an MIC method must be used.
Disk diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
- Inoculum level
- Presence of contamination
- Resistant mutants
- Beta-lactamase activity
- Antagonism and synergy between antibiotics
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Fosfomycin | |
Oxoid Fosfomycin Antimicrobial Susceptibility Disks | |
Antimicrobial Susceptibility Discs | |
5 Cartridges/Pk. | |
50 Discs/Cartridge |
200μg | |
FOT | |
Antimicrobial Susceptibility | |
50 Disks/Cartridge |