missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Half Fraser Supplement

The Oxoid Half Fraser Supplement is used for the enrichment of Listeria species.

£108.00 - £249.00

Specifications

Species Listeria
Description Half Fraser Supplement
Form Freeze-dried
Product Type Microbiology Media
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13235399
View Documents
Thermo Scientific™
SR0166G
For 2.25L of base
£249.00
Pack of 10
In Stock
to see stock.
Add to basket
 
11484209
View Documents
Thermo Scientific™
SR0166E
10/Pk.
£108.00
Pack of 10
In Stock
to see stock.
Add to basket
 
Description

Description

Oxoid Half Fraser Supplement is used for the enrichment of Listeria species.

  • Add to Fraser Broth Base, Part No. CM0895B
Specifications

Specifications

Listeria
Half Fraser Supplement
Freeze-dried
Microbiology Media
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.