Learn More
Thermo Scientific™ Oxoid™ Horse Serum
Thermo Scientific™ Oxoid Horse Serum is an extract of horse blood for addition to culture media.
Brand: Thermo Scientific™ SR0035C
36.76 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 65
Additional Details : Weight : 0.14400kg
Description
Thermo Scientific™ Oxoid Horse Serum is an extract of horse blood for addition to culture media.
Specifications
Horse | |
Liquid | |
Horse Whole Blood, Supplement | |
100 mL |