Learn More
Thermo Scientific™ IMAGEN™ Respiratory Virus Screen Kit using Direct Immunofluorescence Assay
Presumptively detect respiratory syncytical virus (RSV), influenza A and B virus, parainfluenza virus 1,2 & 3, and adenovirus in respiratory specimens with this assay.
Brand: Thermo Scientific™ K612011-2
812.63 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Presumptively detect respiratory syncytical virus (RSV), influenza A and B virus, parainfluenza virus 1,2 & 3 and adenovirus in respiratory specimens with Thermo Scientific™ IMAGEN™ Respiratory Virus Screen Kit. The assay cannot differentiate between the viruses. Individual viruses should be further identified and confirmed using monospecific FITC labeled monoclonal antibody reagents, such as the IMAGEN range, or other methods. The direct respiratory specimen results must be confirmed by cell culture.
Respiratory virus infections are associated with major outbreaks of respiratory disease throughout the world which have a significant impact on world health1,2,3. Viruses cause disease in all age groups but infections are most severe in infant, elderly and immunocompromised populations leading to hospitalization of patients4. Rapid diagnosis of respiratory virus infections is an important aid in the management of patients, prevention, and control of outbreaks, and in influencing the use of antiviral therapy, particularly for influenza A and RSV viral infections.3
Use IMAGEN Respiratory Virus Screen Kit as a rapid, sensitive, and specific method for the presumptive detection of respiratory syncytical virus (RSV), influenza A and B virus, parainfluenza virus 1,2 & 3 and adenovirus in respiratory specimens.
- Rapid results—within a little over 30 minutes of receiving test sample or culture
- Simple—ready-to-use reagents plus all IMAGEN assays follow the same basic methodology
- Excellent sensitivity and specificity—high quality, well-characterized antibodies used in the assay provide excellent sensitivity and specificity
- Easy-to-read results
- Long shelf life—24 months
The viruses mainly responsible for lower respiratory tract infection include RSV, influenza A and B viruses, parainfluenza virus 1, 2 and 3 and adenoviruses. Expected prevalence rates for individual viruses are detailed (with references) in Section 13, Expected Values.
RSV and parainfluenza viruses occur seasonally and are major causes of lower respiratory tract disease in infants and young children. They frequently cause bronchiolitis, croup, bronchitis, and occasionally pneumonia5,6,7. Influenza A and B viruses cause worldwide seasonal epidemics of respiratory disease in adults and infants with a disease spectrum ranging from mild upper respiratory tract symptoms to severe pneumonia8,9. Acute pneumonia in elderly or immunocompromised patients can be life-threatening, particularly when associated with secondary microbial infections.
Adenovirus infections are associated with respiratory, ocular, and enteric disease10. Adenoviruses are reported to be responsible for 5% of acute respiratory diseases in children under the age of 4 years and are a common cause of pharyngitis in young children11.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
IMAGEN™ Respiratory Screen | |
Nasopharyngeal | |
100 Tests |
Respiratory Syncytial Virus (RSV) | |
Reagent |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3712442 | Certificate of Analysis | 02/12/2023 | K612011-2 |
3684810 | Certificate of Analysis | 19/08/2023 | K612011-2 |
3611147 | Certificate of Analysis | 22/05/2023 | K612011-2 |
3440140 | Certificate of Analysis | 22/09/2022 | K612011-2 |
3434610 | Certificate of Analysis | 06/07/2022 | K612011-2 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.