Learn More
Thermo Scientific™ Oxoid™ Kanamycin Sulphate Supplement
Oxoid Kanamycin Sulphate Supplement (10mg/vial) is added to Kanamycin Esculin Azide Agar Base, Part No. CM0591B.
Brand: Thermo Scientific™ SR0092E
54.65 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 65
Additional Details : Weight : 0.17200kg
Description
Oxoid Kanamycin Sulphate Supplement (10mg/vial) is added to Kanamycin Esculin Azide Agar Base, Part No. CM0591B.
Add to Kanamycin Esculin Azide Agar Base, Part No. CM0591BSpecifications
Kanamycin Sulphate Supplement | |
Microbiology Media | |
Each vial for 500mL medium |
Freeze-dried | |
10/Pk. |