missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lab-Lemco Agar (Dehydrated)

Product Code. 10321543
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10321543

Brand: Thermo Scientific™ CM0017B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Examine water and dairy products with this nutritious, general purpose medium.

Examine water and dairy products with nutritious Thermo Scientific™ Oxoid™ Lab-Lemco Agar (Dehydrated)1,2. It is a general-purpose medium for general bacteriological use, and may also be used as the basis for selective, differential, or enriched media. The absence of sodium chloride in the formulation prevents Proteus mirabilis from swarming colonies.

Use Lab-Lamco Agar as a general purpose medium for bacteriological purposes.

It can also be used as nutrient meat extract agar for general bacteriology and for the preservation of stock cultures.

Not all products are available for sale in all territories. Please inquire. 
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Lab-Lemco Agar (Nutrient Agar, U.S. Formulation)
Form Powder
Product Type Agar
Quantity 500 g
Yield For 21.7L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.