Learn More
Thermo Scientific™ Oxoid™ Listeria Selective Enrichment Supplement
Oxoid Listeria Selective Enrichment Supplement is used for the enrichment of Listeria spp.
Brand: Thermo Scientific™ SR0141E
54.25 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 65
Additional Details : CAS Number : 66-81-9 Weight : 0.00050kg Transport : UN number : 2811 Chem class : 6.1 Pack group : II
Description
Oxoid Listeria Selective Enrichment Supplement is used for the enrichment of Listeria spp.
- Add to Listeria Enrichment Broth, Part No. CM0862B
- Each vial supplements 500mL of medium
Specifications
Listeria | |
Freeze-dried | |
10/Pk. |
Listeria Selective Enrichment Supplement | |
Selective Supplement | |
Each vial for 500mL medium |
Safety and Handling
- Listeria Selective Supplement
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
Hazard Statement
- H300-Fatal if swallowed.
- H341-Suspected of causing genetic defects.
- H360-May damage fertility or the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Cycloheximide, Colistin, sulfate, Fosfomycin, Acriflavin Neutral