missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MacConkey Agar (Dehydrated)

Detect, isolate and enumerate coliforms and intestinal pathogens in water, dairy products and biological specimens with this chromogenic medium.

Brand:  Thermo Scientific™ CM0007B

90.97 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10661675

  • £194.00 / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Detect, isolate and enumerate coliforms and intestinal pathogens in water, dairy products and biological specimens with Thermo Scientific™ Oxoid™ MacConkey Agar (Dehydrated). This differential medium gives a number of diagnostic indications such as bile tolerance, colony morphology and chromogenesis. MacConkey Agar corresponds to the medium recommended by the World Health Organizaton1, the Department of Health2 for the bacterial examination of water.

Use MacConkey Agar for the cultivation of pathogens which may be present in a variety of specimens.

  • Easy-to-read: easy to differentiate Escherichia coli (red non-mucoid colonies) from Staphylococcus spp. (pale pink opaque colonies) and Pseudomonas aeruginosa (green-brown) colonies.
  • Recommended by World Health Organizaton1, the Department of Health2.

Due to its ability to support the growth of pathogenic Gram-positive cocci (e.g. staphylococci and enterococci) as well as Enterobacteriaceae, MacConkey Agar is particularly recommended for the cultivation of pathogens which may be present in a variety of specimens such as urine, feces and wound swabs. While it is selective it does not suppress a mixed bacterial flora to the same extent as other inhibitory media (including other MacConkey agars). MacConkey Agar should be used in parallel with other selective indicator media such as Desoxycholate Citrate Agar, Bismuth Sulphite Agar, Brilliant Green Agar and Brilliant Green Bile (2%) Broth, and a non-selective medium such as Blood Agar.

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY
Specifications

Specifications

MacConkey Agar
Agar
For 9.6L medium
Powder
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6233015 Certificate TypeCertificate of Analysis Date06/03/2025 Catalog NumberCM0007B
Lot Number6216184 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberCM0007B
Lot Number6160345 Certificate TypeCertificate of Analysis Date31/08/2024 Catalog NumberCM0007B
Lot Number6130180 Certificate TypeCertificate of Analysis Date05/07/2024 Catalog NumberCM0007B
Lot Number3807409 Certificate TypeCertificate of Analysis Date01/04/2024 Catalog NumberCM0007B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ MacConkey Agar (Dehydrated) > MacConkey Agar; 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.