missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Malt Extract Agar (Dehydrated)

Product Code. 10321063
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10321063

Brand: Thermo Scientific™ CM0059B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect, isolate and enumerate yeasts and molds with typical morphology and pigmentation using medium.

Detect, isolate and enumerate yeasts and molds with Thermo Scientific™ Oxoid™ Malt Extract Agar (Dehydrated). Mycological peptone in the medium gives a luxuriant growth of yeasts and molds with typical morphology and pigmentation.

Malt Extract Agar is similar to the formula of Galloway and Burgess1 used for the detection, isolation, and enumeration of yeasts and molds. For mycological counts, it may be desirable to prepare the more acid medium in order to suppress bacterial growth.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Malt Extract Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 10.0L medium
compliance-icons
Product Identifier
  • MALT EXCTRACT AGAR
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Barley, malt

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.