missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories

Simplify the detection of Gram-negative bacteria with convenient Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories.

Brand:  Thermo Scientific™ MB1093A

19.45 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 13245769

  • £20.15 / 50mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Simplify the detection of Gram-negative bacteria with convenient Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories.

Gram-negative bacteria are of significant concern in both clinical infections and food contamination. Identification of the causative agent is crucial in the clinical setting to identify the cause of infection and to establish an appropriate treatment regime. In the food industry, biochemical identification can help to trace the source of contamination. With these GNB reagents and accessories, Enterobacteriaceae and common miscellaneous Gram-negative bacilli are quickly and accurately identified so that proper, fast treatment for the patient may follow.

  • Fast: More results available overnight
  • Simple: Simple test strip or microplate format
  • Easy to Read and Interpret: Results are clearly visible as distinct color reactions that can be interpreted using the Microbact Identification Package (MB1244A)
  • Comprehensive: Identifies >100 Gram-negative bacteria, more unusual Enterobacteriaceae and oxidase-positive, Gram-negative bacteria
  • Flexible: Use single strips or combine systems for more complex identifications. Ideal for use in clinical or food microbiology laboratories
  • Reliable: A comprehensive range of biochemical tests based on published reference methodologies1-4

Additional items required: Mineral Oil (MB1093A), Oxidase Strips (MB0266A), Microbact Identification Package (Windows™) (MB1244A), Indole Reagent (MB0209A), VPI Reagent (MB0181A), VPII Reagent (MB0184A), TDA Reagent (MB0180A), NIT A Reagent (MB0186A), NIT B Reagent (MB0187A). The 6 reagents above are also available as Reagent Set D (MB1082A).

Not all products are available for sale in all territories. Please inquire.

Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Microbact Mineral Oil
Bottle
1 Box
1 Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories > 50mL/Bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.