missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Modified Preston Campylobacter Selective Supplement
Used for the isolation of Campylobacter jejuni and Campylobacter coli from human, animal, avian, and environmental samples.
Brand: Thermo Scientific™ SR0204E
115.32 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Oxoid Modified Preston Campylobacter Selective Supplement is for the isolation of Campylobacter jejuni and Campylobacter coli from human, animal, avian, and environmental samples.
- Alternative supplement containing amphotericin B as a replacement for cycloheximide
- Add to Campylobacter Agar Base, Part No. CM0689B or Nutrient Broth No. 2, Part No. CM0067B
- Each vial supplements 500mL of medium
Specifications
Campylobacter | |
Freeze-dried | |
10/Pk. |
Modified Preston Campylobacter Selective Supplement | |
Selective Supplement | |
Each vial for 500mL medium |