Learn More
Thermo Scientific™ MRS Broth (Dehydrated)
Isolate and enumerate lactic acid bacteria from food with this non-selective medium.
Brand: Thermo Scientific™ CM0359B
113.55 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 64
Additional Details : Weight : 0.53460kg
Description
Isolate and enumerate lactic acid bacteria from food with Thermo Scientific™ Oxoid™ MRS Agar (DeMan, Rogosa and Sharpe) (Dehydrated). This non-selective medium is designed to support profuse growth of lactic acid bacteria.
Use MRS Agar for enumeration and isolation of mesophilic lactic acid bacteria from food samples. MRS Broth may also be used for tests in the identification of lactobacilli, such as temperature dependence, growth in 4% NaCl, growth in 0.4% Teepol, etc. as recommended by Sharpe, Fryer, and Smith1.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Industrial Reference: AFNOR, IDF, NMKL. | |
Powder | |
500 g |
MRS Broth | |
Dehydrated Culture Media | |
For 9.6L medium |