Learn More
Thermo Scientific™ Oxoid™ Nitrocefin Solution
The Thermo Scientific™ Oxoid Nitrocefin is used for detection of beta-lactamase production.
Brand: Thermo Scientific™ SR0112C
Code : 65
Additional Details : Weight : 0.00100kg Transport : UN number : 1325 Chem class : 4.1 Pack group : II
Description
Thermo Scientific™ Oxoid Nitrocefin is used for detection of beta-lactamase production.
- Reconstitutes to 500μg/mL
- Contents: 5 vials lyophilized nitrocefin (1mg) and 5 vials rehydrating fluid (1.9mL)
Specifications
Nitrocefin | |
5 vials lyophilized nitrocefin (1mg) and 5 vials rehydrating fluid (1.9mL) | |
10 Vial Set/Pk. |
Vial | |
2 x 5 Vials |