missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Novobiocin Selective Supplement
Click to view available options
Quantity:
10 x 10 mg
Unit Size:
Pack of 10
Description
Oxoid Novobiocin Selective Supplement (10mg/vial) is added to EC Broth, Part No. CM0990B; Muller-Kauffmann Tetrathionate-Novobiocin Broth Base, Part No. CM0343B; Brilliant Green Agar, Part No. CM0263B; MRSV, Part No. CM0910B; Modified Tryptone Soya Broth, Part No. CM0989B; and Lysine Iron Agar, Modified, Part No. CM0886B.
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Novobiocin Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10 x 10 mg |
| Yield | Yield will be dependent upon formulation; follow the guidelines for your specific product. |
Safety and Handling
Product Identifier
- Novobiocin Selective Supplement
- Warning
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : novabiocin
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction