missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ ONPG Disc
Thermo Scientific™ Oxoid ONPG Disc is used for rapid detection of beta-galactosidase activity.
Brand: Thermo Scientific™ DD0013T
9.98 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Thermo Scientific™ Oxoid ONPG Disc is used for rapid detection of beta-galactosidase activity.

Specifications
2°C to 8°C | |
ONPG Disk | |
50 Discs |
Disk | |
ONPG Disk |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Oxoid™ ONPG Disc > Remel Oxoid ONPG Disk
Spot an opportunity for improvement?Share a Content Correction