Learn More
Thermo Scientific™ Oxoid™ PBP2' Latex Agglutination Test Kit
For detection of PBP2' and identification of methicillin resistance in Staphylococcus aureus (MRSA) and coagulase-negative Staphylococci.
Brand: Thermo Scientific™ DR0900A
412.95 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 64
Additional Details : Weight : 0.50000kg
Description
Thermo Scientific™ Oxoid PBP2' Latex Agglutination Test is a rapid latex slide agglutination test for detection of PBP2' as an aid in identifying methicillin resistance in both Staphylococcus aureus (MRSA) and coagulase-negative Staphylococci.
Ordering Alerts:
50 tests/kit.
Specifications
Latex Agglutination Test Kit | |
Healthcare | |
50 Tests/Kit |