Learn More
Thermo Scientific™ Oxoid™ Perfringens TSC Supplement
Oxoid Perfringens TSC Supplement (D-cycloserine 200mg/vial) is used for the recovery of Clostridium perfringens.
Brand: Thermo Scientific™ SR0088E
123.68 GBP valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Code : 65
Additional Details : Weight : 0.18000kg
Description
Oxoid Perfringens TSC Supplement (D-cycloserine 200mg/vial) is used for the recovery of Clostridium perfringens.
- Add to DCLS Agar, Part No. CM0393B, or Perfringens Agar Base, TSC, Part No. CM0587B
- Each vial supplements 500mL of medium
Specifications
Perfringens | |
Freeze-dried | |
10/Pk. |
Perfringens TSC Supplement | |
Microbiology Media | |
Each vial for 500mL medium |