Learn More
Thermo Scientific™ Plate Count Agar (Dehydrated)
Determine the plate counts of micro-organisms in food, dairy samples, waste water and other material of sanitary importance with this medium.
Brand: Thermo Scientific™ CM0325B
105.83 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Determine the plate counts of microorganisms in food, dairy samples, waste water and other material of sanitary importance with Thermo Scientific™ Oxoid™ Plate Count Agar (Dehydrated). Plate Count Agar was developed by Buchbinder et al1 which is recommended by APHA2, AOAC3 and PHLS4. The quality control of Plate Count Agar (APHA) includes testing in accordance with ISO 11133:2014.5
Plate Count Agar is equivalent to the medium recommended by the APHA1, PHLS 2 and ISO3,4 for the plate count of micro-organisms in food.
Specifications
Industrial Reference: NMKL. | |
Powder | |
500 g |
Plate Count Agar | |
Agar | |
For 28.6L medium |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Laboratory Use Only