Learn More
Thermo Scientific™ Potato Dextrose Agar (EP/USP/JP/BP) (Dehydrated)
Isolate and enumerate yeasts and molds from dairy and food products with this medium while conforming to EP/USP/JP Microbial Limit Test standards.
Brand: Thermo Scientific™ CM0139B
168.89 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Isolate and enumerate yeasts and molds from dairy and food products1 and prepare Aspergillus niger for the Harmonised Microbial Limit Tests from international pharmacopoeia2,3,4 with Thermo Scientific™ Oxoid™ Potato Dextrose Agar (Dehydrated). This medium is suitable for the detection and enumeration of heat resistant molds in thermally processed fruits and fruit products5.
Pigment production is a critical part of the identification of the fungus. The agar used in the medium is carefully screened to ensure correct pigment production by fungi such as Fusaria spp.
Use Potato Dextrose Agar for detection and enumeration of yeasts and molds in foods.
- Suitable for detection of heat resistant molds in thermally processed fruits and fruit products.
- Suitable medium for the preparation of Aspergillus niger
- Conforms to EP/USP/JP Microbial Limit Test standards2,3,4
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Industrial Reference: FDA. | |
Powder | |
500 g |
Potato Dextrose Agar | |
Agar | |
For 12.8L medium |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6199955 | Certificate of Analysis | 12/12/2024 | CM0139B |
6174073 | Certificate of Analysis | 21/10/2024 | CM0139B |
6160831 | Certificate of Analysis | 24/09/2024 | CM0139B |
6154815 | Certificate of Analysis | 20/08/2024 | CM0139B |
6139989 | Certificate of Analysis | 31/07/2024 | CM0139B |
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24154Expires: 2025-02-28
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Laboratory Use Only