missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Prepared Brilliant Green Bile Broth D/T Bottled Media
Click to view available options
Fill Weight:
10.0-10.3g
Format:
Universal bottle
Unit Size:
240mL
Description
For the detection of coliforms in water and other samples.
Specifications
Specifications
| Color | Green |
| Content And Storage | 2 to 25°C |
| Format | Universal bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Broth |
| Fill Weight | 10.0-10.3g |
| Formulation | Peptone (10.0 g/L); Lactose (10.0 g/L); Ox Bile, purified (20.0 g /L); Brilliant Green (0.0133 g/L) |
| Packaging Type | 24 universal bottles with screw cap closures in a box. |
| Quantity | 24 x 10 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction