missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Prepared Plate Count Agar Bottled Media
Brand: Thermo Scientific™ BO0195T
69.41 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
Straw 1 | |
Sirop w/Screw Cap | |
10 x 250 mL |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4498538 | Certificate of Analysis | 18/03/2025 | BO0195T |
4495044 | Certificate of Analysis | 04/03/2025 | BO0195T |
4493986 | Certificate of Analysis | 25/02/2025 | BO0195T |
4491512 | Certificate of Analysis | 18/02/2025 | BO0195T |
4489173 | Certificate of Analysis | 11/02/2025 | BO0195T |
Special Offers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
0Hours
16Minutes
38Seconds
1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Oxoid™ Prepared Plate Count Agar Bottled Media > Volume: 250mL
Spot an opportunity for improvement?Share a Content Correction