missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Raka-Ray Agar Base (Dehydrated)

Product Code. 10118042
Click to view available options
Description:
Raka-Ray Agar Base
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10118042

Brand: Thermo Scientific™ CM0777B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate lactic acid bacteria in beer and brewing processes with medium as recommended by ASBC and EBC.

Isolate lactic acid bacteria in beer and brewing processes with Thermo Scientific™ Oxoid™ Raka-Ray Agar Base (Dehydrated). The medium is based on the formula of Saha, Sondag and Middlekauff1 and is recommended by the American Society of Brewing Chemists (ASBC)2, and the European Brewing Convention (EBC)3.

Members of the family Lactobacillaceae occurring in the brewing process are important spoilage organisms because products arising from their growth and metabolism are often seriously detrimental to flavor. Detection is complicated by the diverse nutritional and environmental requirements of the family.

Raka-Ray Agar1 enables brewers to monitor in-process beer quickly and accurately for a wide range of organisms including pediococci.

Investigations in which various combinations of growth stimulating agents were added to Universal Beer Agar led to the recognition of a number of agents including sorbitan mono-oleate, liver extract, yeast extract and N-acetyl glucosamine which gave superior results in respect of colony size, colony numbers and incubation time when compared with unmodified Universal Beer Agar.

These investigations provided the basis for the formula of Raka-Ray3 Medium in which sorbitan mono-oleate is included as a stimulant for lactic acid bacteria in general4. Fructose is present as the essential carbohydrate source for Lactobacillus fructivorans5 while maltose is present to detect lactobacilli which cannot utilize glucose6.

TRUSTED_SUSTAINABILITY

Specifications

Description Raka-Ray Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 6.5L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.