Learn More
Thermo Scientific™ Rappaport-Vassiliadis Enrichment Broth (Dehydrated)
Selectively enrich Salmonella spp. from food and environmental specimens with this selective medium.
Brand: Thermo Scientific™ CM0669B
64.07 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 64
Additional Details : Weight : 0.50000kg
Description
Selectively enrich Salmonella spp. from food and environmental specimens with Thermo Scientific™ Oxoid™ Rappaport-Vassiliadis Enrichment Broth (Dehydrated). Rappaport-Vassiliadis Enrichment Broth selects for Salmonella species by exploiting their characteristics when compared with other Enterobacteriaceae, such as the ability to survive at relatively high osmotic pressure, multiply at relatively low pH values, be relatively more resistant to malachite green, and have relatively less demanding nutritional requirements1.
Salmonella are a group of closely related rod-shaped, Gram-negative bacteria. Salmonella typically live in animal and human intestines and are shed through feces. Humans become infected most frequently through contaminated water or food. The most common symptoms of Salmonella infection are diarrhea, fever, abdominal cramps, and vomiting.
Use Rappaport-Vassiliadis Enrichment Broth for selective enrichment of Salmonella species food and environmental samples.
- Selective: malachite green, low pH, and high magnesium chloride concentration inhibit many Gram-positive bacteria, while selectively enriching for salmonellae.
- Superior performance: superior performance to Selenite Enrichment Broth and Tetrathionate Broth for the enrichment of salmonellae with the exception of Salmonella Typhi1.
Rappaport-Vassiliadis (RV) Enrichment Broth is similar to that described by Vassiliadis et al.3 except that the peptone used is soya peptone, which has been reported to enhance the growth of salmonellae1,2.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Rappaport-Vassiliadis Enrichment Broth | |
Dehydrated Culture Media | |
For 16.7L medium |
Powder | |
500 g |
For Laboratory Use Only