Learn More
Thermo Scientific™ Oxoid™ Streptococcal Grouping Disposable Reaction Card
Oxoid Streptococcal Grouping Disposable Reaction Card is used with Streptococcal Grouping Reagents.
Brand: Thermo Scientific™ DR0500G
21.15 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 65
Additional Details : Weight : 0.56700kg
Description
A latex agglutination test for the identification of streptococcal groups A, B, C, D, F, and G.
Specifications
Freeze-dried 2° to 25°C and reconstituted at 2° to 8°C | |
Reaction Card for Streptococcus Testing | |
50 Cards |
Streptococcal Grouping Disposable Reaction Card | |
Group A, B, C, D, F and G |