Learn More
Thermo Scientific™ Oxoid™ Tobramycin Antimicrobial Susceptibility discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
£26.55 - £43.40
Specifications
Antimicrobial Agents Tobramycin
Description Oxoid Tobramycin Antimicrobial Susceptibility Discs
Antibiotic Code TOB
Product Type Antimicrobial Susceptibility Discs
Test Type Antimicrobial Susceptibility
View More Specs
Products 2
Product Code Brand Description Antibiotic Code Concentration Price Quantity & Availability  
Product Code Brand Description Antibiotic Code Concentration Price Quantity & Availability  
11973872
View Documents Promotion Details
Thermo Scientific™
CT0056B
Oxoid Tobramycin Antimicrobial Susceptibility Discs TOB 10µg
£43.40
Pack of 250
Estimated Shipment: 10-01-2025
to see stock.
Edge
Add to basket
11429848
View Documents
Thermo Scientific™
CT1618B
Oxoid Tobramycin Antimicrobial Susceptibility Discs TOB 30μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Tobramycin Antimicrobial Susceptibility discs.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
Tobramycin
TOB
Antimicrobial Susceptibility
50 Discs/Cartridge
6 mm
Oxoid Tobramycin Antimicrobial Susceptibility Discs
Antimicrobial Susceptibility Discs
5 Cartridges/Pk.
50 Discs/Cartridge