Learn More
Thermo Scientific™ Tryptone Soya Agar EP/USP/JP/BP (Dehydrated)
Grow a wide variety of microorganisms with this general-purpose medium while conforming to international pharmacopeia.
Brand: Thermo Scientific™ CM0131R
469.66 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 64
Additional Details : Weight : 3.14000kg
Description
Grow a wide variety of microorganisms with Thermo Scientific ™ Oxoid ™ Tryptone Soya Agar EP/USP/JP/BP (Dehydrated). A general purpose agar medium containing two peptones, supports the growth of both aerobes and anaerobes, the latter being grown either in deep cultures or by incubation under anaerobic conditions. Tryptone Soya Agar is recommended as a reference medium when testing selective media to measure the degree of inhibition1.
Tryptone Soya Agar, also known as Casein Soya Bean Digest Agar, is a general purpose medium specified in pharmacopeia and food testing methods for the growth of a wide variety of organisms. Recommended as a reference medium for testing selective media. Conforms to formulations detailed in the international pharmacopoeia2,3,4,5.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Tryptone Soya Agar | |
Agar | |
For 62.5L medium |
Powder | |
2.5 kg |
For Laboratory Use Only