Learn More
Thermo Scientific™ Oxoid™ Tryptone
The Thermo Scientific™ Oxoid Tryptone ia a pancreatic digest of casein for use where high performance and uniformity of composition are of paramount importance.
Brand: Thermo Scientific™ LP0042R
556.83 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 64
Additional Details : Weight : 0.30000kg
Description
The Thermo Scientific™ Oxoid Tryptone ia a pancreatic digest of casein for use where high performance and uniformity of composition are of paramount importance.
Specifications
Tryptone | |
Powder | |
Microbiology Media | |
1 Each |