missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thioglycollate Medium USP (Dehydrated)

Cultivate aerobic and anaerobic organisms in the performance of sterility tests with this medium.

Brand:  Thermo Scientific™ CM0173B

83.74 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10321003

  • £170.00 / 500g
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Cultivate aerobic and anaerobic organisms in the performance of sterility tests with Thermo Scientific™ Oxoid™ Thioglycollate Medium USP (Dehydrated). The medium is prepared according to the formula specified in the US Pharmacopoeia1 for the performance of sterility tests and conforms to formulations detailed in the British and European Pharmacopoeia 2,3,4.

Thioglycollate Medium is suitable for the cultivation of both aerobic and anaerobic organisms. Sodium thioglycollate acts as a reducing agent and neutralizes the toxic effects of mercurial compounds used as preservatives in solutions for injection, etc.

  • Well-buffered medium ensures that acid or alkaline inocula produce negligible alteration in the reaction of the medium
  • Suitable for cultivation of both aerobic and anaerobic organisms.

Not all products are available for sale in all territories. Please inquire.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Thioglycollate Medium, USP
Dehydrated Culture Media
For 16.9L medium
Powder
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Laboratory Use Only