Learn More
Thermo Scientific™ Oxoid™ Vancomycin Supplement
Used for addition to selective media for isolation of Vancomycin Resistant Enterococci and High Level Aminoglycoside Resistant Enterococci from clinical specimens.
Brand:  Thermo Scientific™ SR0247E
98.55 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12638797
  • £148.00 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Oxoid Vancomycin Supplement is for addition to selective media for the isolation of Vancomycin Resistant Enterococci (VRE) and High Level Aminoglycoside Resistant Enterococci (HLARE) from clinical specimens.
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vancomycin-resistant Enterococci (VRE), High-level Aminoglucoside-resistant Enterococci (HLARE)
Powder
10/Pk.
Oxoid™ Vancomycin Supplement
Microbiology Media