missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Violet Red Bile Agar
GREENER_CHOICE

Product Code. 13225609
Click to view available options
Quantity:
500 g
2.5kg
Unit Size:
3kg
500g
This item is not returnable. View return policy

Product Code. 13225609

Brand: Thermo Scientific™ CM0107R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect and enumerate coliform organisms in water, food and dairy products with this selective medium.

Detect and enumerate coliform organisms in water, food and dairy products with Thermo Scientific™ Oxoid™ Violet Red Bile Agar (Dehydrated). This lactose containing selective medium has been used for the determination of the coli-aerogenes content of water, milk and other dairy products, dairy equipment, and food products1,2. Due to the addition of selective inhibitory components, neutral red, crystal violet, and bile salts, the medium provides selectivity against non-target Gram-positive bacteria.

Use Violet Red Bile Agar for examination of food, animal feed, and environmental samples for the presence of indicator groups such as coliforms. Besides safety, indicator groups provide a measure of food quality and spoilage potential. One of the most common groups of bacteria employed as indicator organisms by the food industry are the coliforms, which can be regarded as a subgroup within the Enterobacteriaceae.

  • Easy-to-read: organisms rapidly fermenting the lactose (coliforms) form red-purple colonies with red-purple halos; while late lactose fermenters form pale colonies with greenish halos.
  • Selective: crystal violet and bile salts suppress the growth of non-target Gram-positive bacteria.

Druce et al.3 found that Violet Red Bile Lactose Agar was as good an indicator of coli-aerogenes bacteria in milk as MacConkey Broth, and that the Violet Red Bile Agar was suitable for determining the coli-aerogenes content of milk.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Violet Red Bile Agar
Form Powder
Product Type Agar
Quantity 2.5kg
Yield For 13.0 L mediμm

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.