Learn More
Thermo Scientific™ Oxoid™ Vitox Supplement
Oxoid Vitox Supplement is used for the preparation of Thayer Martin Medium and New York City Medium for the isolation of Neisseria spp.
Brand: Thermo Scientific™ SR0090A
170.54 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 65
Additional Details : Weight : 2.72160kg
Description
Oxoid Vitox Supplement is used for the preparation of Thayer Martin Medium and New York City Medium for the isolation of Neisseria spp.
- Contents: Vitox Supplement (5 vials) plus reconstitution fluid (5 vials)
- Add to GC Agar Base, Part No. CM0367B
Ordering Information:
- Each vial of Vitox plus reconstitution fluid supplements 500mL of medium
Includes:
- Five vials of Vitox plus five vials of reconstitution fluid
Specifications
Vitox Supplement | |
Microbiology Media | |
5 x Freeze dried vial + 5 x Rehydration fluid |
Freeze-dried | |
2 x 5/Pk. | |
Each vial for 500mL medium |