Learn More
Thermo Scientific™ Yeast Extract Agar (Dehydrated)
Carry out plate count of microorganisms in water with this medium while adhering to ISO 1133:2014.
Brand: Thermo Scientific™ CM0019B
124.50 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 64
Additional Details : Weight : 0.53640kg
Description
Carry out plate counts of microorganisms in water with Thermo Scientific™ Oxoid™ Yeast Extract Agar (Dehydrated). Yeast Extract Agar is made to the formula described by Windle Taylor1 for the plate count of microorganisms in water and is tested in accordance with ISO 1133:20142. It is a rich, nutritious medium for the recovery of a wide range of bacteria, yeast, and molds.
The usual method of counting heterotrophic bacteria in water is by the poured plate method with Yeast Extract Agar. Separate counts are made of those aerobic mesophilic organisms which form visible colonies in this medium after 24 hours incubation at 37°C and those which form colonies after three days incubation at 22°C. The two methods give different results2.
The organisms growing under these conditions comprise bacteria, yeasts, and molds.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Yeast Extract Agar | |
Agar | |
For 21.7L medium |
Powder | |
500 g |
For Laboratory Use Only