missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Yeast Extract Powder

Product Code. p-4522648
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
Unit Size:
2.5kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10108202

Brand: Thermo Scientific™ LP0021B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxoid Yeast Extract Powder is a dried yeast autolysate of proprietary specification optimized for fermentation and culture media applications.

Thermo Scientific™ Oxoid Yeast Extract Powder is a proprietary dried yeast autolysate which provides a nutritious source of nitrogen and vitamins optimized for use in culture media or fermentation.

Increases yield of organisms in many media or fermentation broths.

  • Recommended for plate count of organisms in water
  • Increases yield of organisms of many media or fermentation broths
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Dehydrated Powder, Ambient & Dry Storage
Description Yeast Extract Powder
Form Dehydrated Powder
Product Type Peptone
For Use With (Application) Culture media or fermentation processes
Quantity 500 g
Protein Source Yeast Extract

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.