missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Yeast and Mold Agar (Dehydrated)

Isolate yeasts and molds within the brewing industry with this medium.

Brand:  Thermo Scientific™ CM0920B

259.74 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11923922

  • £580.00 / 500g
Estimated Shipment: 19-02-2025
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Isolate yeasts and molds with Thermo Scientific™ Oxoid™ Yeast and Mold Agar (Dehydrated). The medium is based on the formulation described by Wickerham1,2 and is formulated specifically for suitability within the brewing industry, for detection of both Saccharomyces and non-saccharomyces wild yeasts in the presence of culture yeasts.

The medium utilizes the different sensitivities of wild yeast vs. culture ale or lager yeast to the inhibitory action of copper. This enables wild yeasts to grow on the medium while suppressing the growth of culture yeast.

The medium is highly nutritious for the growth of yeasts and molds. Peptones and yeast extracts provide nitrogenous compounds and vitamin B complex. Dextrose acts as the energy source. Yeast and Mold Agar is optimized to for addition of copper usually within the range 50-300 mg/L depending on the sensitivity of the culture yeast to copper, as recommended by the Institute of Brewing Methods of analysis3.

The complete medium is recommended for examination for the microbiological quality of beers in process, pitching yeasts and packaged beers.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Yeast and Mold Agar
Agar
For 12.2L medium
Powder
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6108247 Certificate TypeCertificate of Analysis Date02/11/2024 Catalog NumberCM0920B
Lot Number3627651 Certificate TypeCertificate of Analysis Date18/03/2023 Catalog Number
Lot Number3586718 Certificate TypeCertificate of Analysis Date10/12/2022 Catalog Number
Lot Number3543737 Certificate TypeCertificate of Analysis Date07/10/2022 Catalog Number
Lot Number3530930 Certificate TypeCertificate of Analysis Date17/09/2022 Catalog Number
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Yeast and Mold Agar (Dehydrated) > 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use Only