missing translation for 'onlineSavingsMsg'
Learn More

Oxolinic acid, 98%, Thermo Scientific Chemicals

Product Code. 10564408
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14698-29-4
Molecular Formula C13H11NO5
Molecular Weight (g/mol) 261.23
MDL Number MFCD00056775
InChI Key KYGZCKSPAKDVKC-UHFFFAOYSA-N
Synonym oxolinic acid, nidantin, dioxacin, emyrenil, prodoxal, prodoxol, ossian, gramurin, oksaren, oxolinic
PubChem CID 4628
ChEBI CHEBI:138856
IUPAC Name 5-ethyl-8-oxo-[1,3]dioxolo[4,5-g]quinoline-7-carboxylic acid
SMILES CCN1C=C(C(O)=O)C(=O)C2=CC3=C(OCO3)C=C12

Specifications

Melting Point 314.0°C to 316.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 15,7044
Formula Weight 261.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Oxolinic acid
Show More Show Less
compliance-icons
Product Identifier
  • Oxolinic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'