missing translation for 'onlineSavingsMsg'
Learn More

Oxone∣r, monopersulfate, Thermo Scientific Chemicals

£38.69 - £230.15

Chemical Identifiers

CAS 70693-62-8
Molecular Formula H3K5O18S4
Molecular Weight (g/mol) 614.74
MDL Number MFCD00040551
InChI Key HJKYXKSLRZKNSI-UHFFFAOYSA-I
Synonym Potassium peroxymonosulfate, Potassium monopersulfate triple salt
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11443850
View Documents Promotion Details
Thermo Scientific Alfa Aesar
089892.22
100 g
£38.69
100g
Please to purchase this item. Need a web account? Register with us today!
11463850
View Documents
Thermo Scientific Alfa Aesar
089892.A1
1 kg
£66.73
1kg
Please to purchase this item. Need a web account? Register with us today!
11453850
View Documents
Thermo Scientific Alfa Aesar
089892.A7
5 kg
£230.15
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Oxone is used for halogenation of a,b-unsaturated carbonyl compounds and catalytic generation of hypervalent iodine reagents for alcohol oxidation. It is a used for rapid, and good synthesis of oxaziridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

70693-62-8
614.74
HJKYXKSLRZKNSI-UHFFFAOYSA-I
H3K5O18S4
MFCD00040551
Potassium peroxymonosulfate, Potassium monopersulfate triple salt

Specifications

70693-62-8
H3K5O18S4
Potassium peroxymonosulfate, Potassium monopersulfate triple salt
HJKYXKSLRZKNSI-UHFFFAOYSA-I
615.5
1.15 g/mL
Granules
MFCD00040551
Soluble in water (100mg/ml).
614.74
Hygroscopic
Oxone
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302+H332-H314-H317-H334-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 274-778-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Oxone, monopersulfate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Potassium peroxymonosulfate sulfate, Potassium pyrosulfate,Dipotassium peroxodisulphate, Potassium

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.