missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Oxonic acid, potassium salt, 97.5%
£78.00 - £224.00
Chemical Identifiers
CAS | 2207-75-2 |
---|---|
Molecular Formula | C4H2KN3O4 |
Molecular Weight (g/mol) | 195.18 |
MDL Number | MFCD00010565 |
InChI Key | IAPCTXZQXAVYNG-UHFFFAOYSA-M |
Synonym | potassium oxonate, oteracil potassium, oxonic acid potassium salt, allantoxanic acid potassium salt, potassium azaorotate, oxonic acid, potassium salt, oxonate, potassium, oxonate, allantoxanic acid |
PubChem CID | 2723920 |
ChEBI | CHEBI:80230 |
IUPAC Name | potassium;4,6-dioxo-1H-1,3,5-triazine-2-carboxylate |
SMILES | [K+].[O-]C(=O)C1=NC(=O)NC(=O)N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10524111
|
Thermo Scientific Chemicals
168530250 |
25 g | Glass bottle |
£78.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10173750
|
Thermo Scientific Chemicals
168531000 |
100 g | Glass bottle |
£224.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2207-75-2 | |
195.18 | |
IAPCTXZQXAVYNG-UHFFFAOYSA-M | |
2723920 | |
potassium;4,6-dioxo-1H-1,3,5-triazine-2-carboxylate |
C4H2KN3O4 | |
MFCD00010565 | |
potassium oxonate, oteracil potassium, oxonic acid potassium salt, allantoxanic acid potassium salt, potassium azaorotate, oxonic acid, potassium salt, oxonate, potassium, oxonate, allantoxanic acid | |
CHEBI:80230 | |
[K+].[O-]C(=O)C1=NC(=O)NC(=O)N1 |
Specifications
2207-75-2 | |
White | |
97.5% | |
MFCD00010565 | |
potassium oxonate, oteracil potassium, oxonic acid potassium salt, allantoxanic acid potassium salt, potassium azaorotate, oxonic acid, potassium salt, oxonate, potassium, oxonate, allantoxanic acid | |
[K+].[O-]C(=O)C1=NC(=O)NC(=O)N1 | |
195.18 | |
CHEBI:80230 | |
97.5% | |
Oxonic acid, potassium salt, 97.5% |
>300.0°C | |
Authentic | |
C4H2KN3O4 | |
24, 451 | |
IAPCTXZQXAVYNG-UHFFFAOYSA-M | |
potassium;4,6-dioxo-1H-1,3,5-triazine-2-carboxylate | |
2723920 | |
195.18 | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 218-627-5
missing translation for 'rtecsNumber' : RR4580000
RUO – Research Use Only