missing translation for 'onlineSavingsMsg'
Learn More

OxPhos Human WB Antibody Cocktail, Invitrogen™

Antibody Cocktail

Brand:  Invitrogen 458199

Product Code. 10698303

  • £698.00 / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

This product is a cocktail of multiple OxPhos Mouse anti Human antibodies. C-I-20 (ND6) (clone) 20E9DH10C12 - Bovine Complex I This is an optimized premixed cocktail suitable for Western Blotting analysis of the relative levels of the 5 OxPhos complexes in mitochondrial preparations from human sources. These clones were chosen because they are against a subunit that is labile when its complex is not assembled. Moreover, the combination is readily resolved in SDS-PAGE when the appropriate gel conditions are used. C-II-30 (FeS) (clone) 21A11AE7 - Bovine Heart Complex II C-III-Core 2 (clone) 13G12AF12BB11 - Bovine Heart Complex II, III C-IV-II (clone) 12C4F12 -Human Complex IV, subunit II C-V-alpha (clone) 15H4C4 -Bovine Complex V

OxPhos Human WB Cocktail antibody specifically detects OxPhos Human WB in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OxPhos Human WB
Cocktail
Unconjugated
12C4F12; 13G12AF12BB11; 15H4C4; 20E9DH10C12; 21A11AE7; OxPhos Complex
IgG fraction
RUO
Human
Antibody
Ig
Western Blot
1.5 mg/mL
HEPES buffered saline with 0.02% sodium azide
Mouse
360 μg
Primary
4° C, do not freeze
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers