Learn More
Oxygen Cylinder Trolleys
Brand:  N/A TS178L
Encompass_Preferred
Product Code. 12925961
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
The tubular frame has a sheet steel base and retaining straps to hold the cylinder in position. Each trolley is fitted with anti-static metal centred wheels and front 100mm non-marking corner buffers.
Specifications
Specifications
1100 mm
420 mm
Oxygen Cylinder
Oxygen Cylinder
420 x 400 x 1100 mm
400 mm
Oxygen Cylinder Trolleys
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Oxygen Cylinder Trolleys