Learn More
PureAire Monitoring Systems, Inc. Oxygen Deficiency Monitor 0-25% with 10+ Year Sensor
Perfect for facilities that use nitrogen, helium, and argon. The monitor is best suited for confined spaces, cryogenic locations, and freezers. Remains accurate at temperatures as low as -40°C. Our zirconium sensor lasts 10 years or more.
Brand: PureAire Monitoring Systems, Inc.
Additional Details : Weight : 0.61960kg
Description
- Long-life 10+ year average sensor life
- Two-Alarm setpoints for 19.5% and 18.0%
- Built-in audible alarm 90 db
- Digital display, and 4-20mA analog output
- Wall mounting brackets
Why choose PureAire?
- As an industry leader in oxygen monitoring and gas detection, PureAire is dedicated to the safety of our clients, and the customers they serve, by manufacturing monitors that are long-lasting, innovative, and reliable.
What is an O2 Monitor?
- The Air Check O2 Deficiency Monitor is designed for continuous monitoring where low oxygen levels may pose a hazard to personnel.
What gases can an O2 monitor be used for?
- PureAire’s oxygen monitors can be used for nitrogen, argon, and helium.
Installation is easy. Plug-in ready.
- The monitor will arrive ready to work right out of the box. Simply secure the unit on the wall 1-2 meters from the gas source, plug it in, and power it on. The monitor will begin sampling oxygen levels after a 4 minute startup period. Levels are displayed clearly on the screen for easy access and visibility.
Certifications
CE Marked
Services
2 years
Specifications
3 in. | |
5.125 x 3.15 x3 in. | |
80 mm | |
-40°F to +134°F | |
24 VDC / 250mA | |
1.1 lb. | |
5.125 in. |
76 mm | |
3.15 in. | |
0-95% RH | |
-40 to +55°C | |
2 Year after ETD | |
0.5 kg | |
130 mm |