Learn More
PureAire Monitoring Systems, Inc. Oxygen Deficiency Monitor with 3m Cable and 10+ Year Sensor
Perfect for facilities that use nitrogen, helium, and argon. The monitor is best suited for confined spaces, cryogenic locations, and freezers. 3-meter cable included. The monitor remains accurate at temperatures as low as -40°C. Our zirconium sensor lasts 10 years or more.
Brand: PureAire Monitoring Systems, Inc.
Additional Details : Weight : 0.83640kg
Description
- Long-life 10+ year average sensor life
- Two-Alarm setpoints for 19.5% and 18.0%
- Built-in audible alarm 90 db
- 3 meter cable
- Digital display, and 4-20mA analog output
- Wall mounting brackets
Why choose PureAire?
- As an industry leader in oxygen monitoring and gas detection, PureAire is dedicated to the safety of our clients, and the customers they serve, by manufacturing monitors that are long-lasting, innovative, and reliable.
What is an O2 Monitor?
- The Air Check O2 Deficiency Monitor is designed for continuous monitoring where low oxygen levels may pose a hazard to personnel.
Installation is easy. Plug-in ready.
- The monitor will arrive ready to work right out of the box. Simply secure the unit on the wall 1-2 meters from the gas source, and plug it in. The monitor will begin sampling oxygen levels after a 4 minute startup period. Levels are displayed clearly on the screen for easy access and visibility.
Certifications
CE Marked
Services
2 years
Specifications
CE | |
76 mm | |
3.15 in. | |
0-95% | |
-40 to +55°C | |
2 Year after ETD | |
0.5 kg | |
130 mm |
3 in. | |
3 x 5.125 x 3.25 in. | |
80 mm | |
-40°F to +134°F | |
24V | |
1.1 lb. | |
5.125 in. |