Learn More
PureAire Monitoring Systems, Inc. Oxygen Deficiency Monitor KF25 with 10+ Year Sensor
For use in a vacuum environment, OLED printer, nitrogen generator, or machines using nitrogen, and cryogenics. The robust 10 year average life zirconium sensor is connected via the KF25 flange. Sensor is connected to the monitor using a 3m cable for easy connection.
Brand: PureAire Monitoring Systems, Inc.
Additional Details : Weight : 0.90260kg
Description
- Long-life 10+ year average sensor life
- Two-Alarm setpoints for 19.5% and 18.0%
- Built-in audible alarm 90 db
- Digital display, and 4-20mA analog output
- Wall mounting brackets
Why choose PureAire?
- As an industry leader in oxygen monitoring and gas detection, PureAire is dedicated to the safety of our clients, and the customers they serve, by manufacturing monitors that are long-lasting, innovative, and reliable.
What is an O2 Monitor?
- The Air Check O2 Deficiency Monitor is designed for continuous monitoring where low oxygen levels may pose a hazard or a risk to personnel.
What gases can an O2 monitor be used for?
- PureAire’s oxygen monitors can be used for nitrogen, argon, and helium.
Installation is easy. Plug-in ready.
- The monitor will arrive ready to work right out of the box. Simply secure the unit on the wall 1-2 meters from the gas source, plug it in, and power it on. The monitor will begin sampling oxygen levels after a 4 minute startup period. Levels are displayed clearly on the screen for easy access and visibility.
Certifications
CE Marked
Services
2 years
Specifications
3 in. | |
3 x 5.125 x 3.15 in. | |
80 mm | |
-40°F to +134°F | |
24 VDC | |
1.1 lb. | |
5.125 in. |
76 mm | |
3.15 in. | |
0-95% | |
-40 to +55°C | |
2 Year after ETD | |
0.5 kg | |
130 mm |