missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P 22077, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Inhibitor of ubiquitin-specific protease (USP) 7 (EC50 = 8.6 μM); also inhibits the closely related deubiquitinase (DUB) USP47. Demonstrates downstream inhibition of HDM2 and claspin in vitro. Inhibits USP7-mediated p53 deubiquitination. Blocks deubiquitination of Tip60 (KAT5) histone lysine acetyltransferase.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 315.32 |
Percent Purity | >99% |
Chemical Name or Material | P 22077 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'