missing translation for 'onlineSavingsMsg'
Learn More

p-Anisaldehyde, 99+%, Thermo Scientific Chemicals

Product Code. 11309881
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-11-5
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00003385
InChI Key ZRSNZINYAWTAHE-UHFFFAOYSA-N
Synonym p-anisaldehyde, anisaldehyde, anisic aldehyde, p-methoxybenzaldehyde, 4-anisaldehyde, aubepine, p-anisic aldehyde, p-formylanisole, benzaldehyde, 4-methoxy, crategine
PubChem CID 31244
ChEBI CHEBI:28235
IUPAC Name 4-methoxybenzaldehyde
SMILES COC1=CC=C(C=C1)C=O

Specifications

Melting Point -1°C
Color Colorless to Yellow
Density 1.1210g/mL
Boiling Point 248°C to 249°C
Flash Point 116°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4CHO
Refractive Index 1.5710 to 1.574
Quantity 2.5 L
Beilstein 08,67
Merck Index 15,656
Specific Gravity 1.121
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: miscible withe alcohol and ether,soluble in 5 volumes of 50% alcohol,soluble in most organic solvents
Viscosity 4.1 mPa.s (25°C)
Formula Weight 136.15
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material p-Anisaldehyde, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'