missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Anisidine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
1000 g
Description
p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.
Solubility
Soluble in acetone, benzene, ether and ethanol. Insoluble in water.
Notes
Air and light sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and chloroformates.
p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.
Solubility
Soluble in acetone, benzene, ether and ethanol. Insoluble in water.
Notes
Air and light sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and chloroformates.
Specifications
Specifications
Melting Point | 56°C to 60°C |
Density | 1.089 |
Boiling Point | 240°C to 243°C |
Flash Point | 122°C (251°F) |
Quantity | 1000 g |
UN Number | UN2811 |
Beilstein | 471556 |
Merck Index | 14,667 |
Solubility Information | Soluble in acetone,benzene,ether and ethanol. Insoluble in water. |
Formula Weight | 123.16 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'