missing translation for 'onlineSavingsMsg'
Learn More

p-Anisidine, 99%, Thermo Scientific Chemicals

Product Code. 11498690
Click to view available options
Quantity:
100 g
500 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.

Solubility
Soluble in acetone, benzene, ether and ethanol. Insoluble in water.

Notes
Air and light sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-94-9
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007864
InChI Key BHAAPTBBJKJZER-UHFFFAOYSA-N
Synonym p-anisidine, 4-anisidine, 4-methoxybenzenamine, 4-aminoanisole, p-aminoanisole, para-anisidine, p-methoxyaniline, benzenamine, 4-methoxy, 4-methoxybenzeneamine, p-anisylamine
PubChem CID 7732
ChEBI CHEBI:82388
IUPAC Name 4-methoxyaniline
SMILES COC1=CC=C(C=C1)N

Specifications

Melting Point 56°C to 60°C
Density 1.089
Boiling Point 240°C to 243°C
Flash Point 122°C (251°F)
Quantity 500 g
UN Number UN2811
Beilstein 471556
Merck Index 14,667
Solubility Information Soluble in acetone,benzene,ether and ethanol. Insoluble in water.
Formula Weight 123.16
Percent Purity 99%
Chemical Name or Material p-Anisidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'