missing translation for 'onlineSavingsMsg'
Learn More

p-Benzoquinone dioxime, 95%, Thermo Scientific Chemicals

£23.90 - £238.00

Chemical Identifiers

CAS 105-11-3
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00063636
InChI Key LNHURPJLTHSVMU-CGXWXWIYSA-N
Synonym 1,4-benzoquinone dioxime, p-benzoquinonedioxime, p-benzoquinone dioxime, p-quinone dioxime, quinone dioxime, benzoquinone dioxime, p-quinone oxime, actor q, dioxime p-benzoquinone, dibenzo pqd
PubChem CID 7737
IUPAC Name N-(4-nitrosophenyl)hydroxylamine
SMILES ON=C1C=CC(C=C1)=NO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11478157
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A17532.18
50 g
£23.90
50g
Please to purchase this item. Need a web account? Register with us today!
11488157
View Documents
Thermo Scientific Alfa Aesar
A17532.30
250 g
£76.40
250g
Please to purchase this item. Need a web account? Register with us today!
11468157
View Documents
Thermo Scientific Alfa Aesar
A17532.0B
1000 g
£238.00
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

105-11-3
138.13
LNHURPJLTHSVMU-CGXWXWIYSA-N
7737
ON=C1C=CC(C=C1)=NO
C6H6N2O2
MFCD00063636
1,4-benzoquinone dioxime, p-benzoquinonedioxime, p-benzoquinone dioxime, p-quinone dioxime, quinone dioxime, benzoquinone dioxime, p-quinone oxime, actor q, dioxime p-benzoquinone, dibenzo pqd
N-(4-nitrosophenyl)hydroxylamine

Specifications

105-11-3
1.49
MFCD00063636
1,4-benzoquinone dioxime, p-benzoquinonedioxime, p-benzoquinone dioxime, p-quinone dioxime, quinone dioxime, benzoquinone dioxime, p-quinone oxime, actor q, dioxime p-benzoquinone, dibenzo pqd
ON=C1C=CC(C=C1)=NO
138.13
138.13
p-Benzoquinone dioxime
∼223°C (decomposition)
C6H6N2O2
2043234
LNHURPJLTHSVMU-CGXWXWIYSA-N
N-(4-nitrosophenyl)hydroxylamine
7737
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.

P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H351

missing translation for 'einecsNumber' : 203-271-5

missing translation for 'rtecsNumber' : DK4900000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documenti

Documenti

Certificati

RUO – Research Use Only