missing translation for 'onlineSavingsMsg'
Learn More

p-Chloranil, 97%, Thermo Scientific Chemicals

Product Code. 11435593
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

p-Chloranil is used as a dye intermediate, oxidizing agent, vulcanizing agent and dehydrogenation reagent. It is also used to make chloranil electrodes for pH measurements. Further, it serves as a hydrogen acceptor and used for the aromatization reactions such as conversion of cyclohexadienes to the benzene derivatives. It is used to test the secondary amine. In addition to this, it is employed as a precursor to prepare diaziquone, which is used as a cancer chemotherapeutic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Chloranil is used as a dye intermediate, oxidizing agent, vulcanizing agent and dehydrogenation reagent. It is also used to make chloranil electrodes for pH measurements. Further, it serves as a hydrogen acceptor and used for the aromatization reactions such as conversion of cyclohexadienes to the benzene derivatives. It is used to test the secondary amine. In addition to this, it is employed as a precursor to prepare diaziquone, which is used as a cancer chemotherapeutic agent.

Solubility
Soluble in ether and acetone. Slightly soluble in chloroform, dimethylformamide, solvent naphtha, benzene, carbon disulfide and carbon tetrachloride. Insoluble in water, cold petroleum ether, methanol, dibutyl phthalate and cold alcohol.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-75-2
Molecular Formula C6Cl4O2
Molecular Weight (g/mol) 245.86
MDL Number MFCD00001594
InChI Key UGNWTBMOAKPKBL-UHFFFAOYSA-N
Synonym chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone
PubChem CID 8371
ChEBI CHEBI:36703
IUPAC Name 2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione
SMILES ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O

Specifications

Melting Point ∼290°C (sublimation)
Density 1.97
Quantity 100 g
UN Number UN3077
Beilstein 393006
Merck Index 14,2078
Solubility Information Soluble in ether and acetone. Slightly soluble in chloroform,dimethylformamide,solvent naphtha,benzene,carbon disulfide and carbon tetrachloride. Insoluble in water,cold petroleum ether,methanol,dibutyl phthalate and cold alcohol.
Formula Weight 245.88
Percent Purity 97%
Chemical Name or Material p-Chloranil
compliance-icons compliance-icons
Product Identifier
  • p-Chloranil
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : p-Chloranil

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'