missing translation for 'onlineSavingsMsg'
Learn More

p-Cresol, 0.99%, Honeywell™

Catalog Number 15697780
Click to view available options
Quantity:
5 g
100 g
500 g
2 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
106-44-5
C7H8O
108.14
MFCD00002376
IWDCLRJOBJJRNH-UHFFFAOYSA-N
p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
2879
CHEBI:17847
4-methylphenol
CC1=CC=C(C=C1)O
This item is not returnable. View return policy
106-44-5
C7H8O
108.14
MFCD00002376
IWDCLRJOBJJRNH-UHFFFAOYSA-N
p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
2879
CHEBI:17847
4-methylphenol
CC1=CC=C(C=C1)O
Melting Point 32°C to 35°C
Color Colorless
Density 1.034g/mL at 25°C
Boiling Point 202°C
Flash Point 85°C (185°F)
Ignition Point 1038°F
Packaging Glass bottle
Linear Formula CH3C6H4OH
Quantity 2 kg
UN Number UN3455
Beilstein 1305151
Vapor Density 3.72 (vs air)
Formula Weight 108.14g/mol
Percent Purity 0.99
Chemical Name or Material p-Cresol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'