Learn More
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
£24.82 - £141.71
Chemical Identifiers
CAS | 106-44-5 |
---|---|
Molecular Formula | C7H8O |
Molecular Weight (g/mol) | 108.14 |
MDL Number | MFCD00002376 |
InChI Key | IWDCLRJOBJJRNH-UHFFFAOYSA-N |
Synonym | p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene |
PubChem CID | 2879 |
ChEBI | CHEBI:17847 |
IUPAC Name | 4-methylphenol |
SMILES | CC1=CC=C(C=C1)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10790282
|
Thermo Scientific Acros
405740050 |
5 g |
£24.82
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12163741
|
Thermo Scientific Acros
405741000 |
100 g |
£30.65
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10351631
|
Thermo Scientific Acros
405745000 |
500 g |
£38.79
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10701904
|
Thermo Scientific Acros
405740025 |
2.5 kg |
£141.71
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
106-44-5 | |
108.14 | |
IWDCLRJOBJJRNH-UHFFFAOYSA-N | |
2879 | |
4-methylphenol |
C7H8O | |
MFCD00002376 | |
p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene | |
CHEBI:17847 | |
CC1=CC=C(C=C1)O |
Specifications
106-44-5 | |
Colorless to Yellow | |
202.0°C | |
Authentic | |
Glass bottle | |
CH3C6H4OH | |
06,389 | |
1.034 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether and chloroform | |
CC1=CC=C(C=C1)O | |
108.14 | |
CHEBI:17847 | |
108.14 | |
Pure | |
p-Cresol, 99+% |
32.0°C to 34.0°C | |
1.0340g/mL | |
89°C | |
99% min. (GC) | |
C7H8O | |
MFCD00002376 | |
15,2564 | |
p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene | |
IWDCLRJOBJJRNH-UHFFFAOYSA-N | |
4-methylphenol | |
2879 | |
8 mPa.s (35°C) | |
99+% | |
Crystalline Solid or Liquid |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-398-6
missing translation for 'rtecsNumber' : GO6475000
missing translation for 'tsca' : TSCA
- p-Cresol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H412-Harmful to aquatic life with long lasting effects.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only